Please login to your online account to display your discounted pricing

Mouse anti-Cytokeratin, Basic (Type II or HMW), Clone: SPM116, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23439501MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23439501

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Bovine
Cross Reactivity Canine
Cross Reactivity Chicken
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Cross Reactivity Rabbit
Cross Reactivity Rat
Format Purified
Quantity 0.1mg
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Immunogen Solubilized keratin extract from human stratum corneum
Host Species Mouse
Antigen Cytokeratin, Basic (Type II or HMW)
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Monoclonal or Polyclonal Monoclonal
Regulatory Status RUO
Purification Method Purified
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Clone SPM116
Concentration 1mg/mL
Reconstitution Protein A purified
Test Specificity This MAb recognizes basic (Type II or HMW) cytokeratins, which include 67kDa (CK1); 64kDa (CK3); 59kDa (CK4); 58kDa (CK5); 56kDa (CK6); 52kDa (CK8). This MAb recognizes the 65-67, 64, 59, 58, 56, and 52kDa keratins of basic subfamily.
Isotype IgG1
Primary or Secondary Primary
Conjugate Unlabeled