Please login to your online account to display your discounted pricing

Mouse anti-Cytokeratin, Acidic (Type I or LMW), Clone: SPM115, Novus Biologicals™

$139.00 - $321.00

Specifications

Antigen Cytokeratin, Acidic (Type I or LMW)
Clone SPM115
Concentration 0.2mg/mL
Immunogen Solubilized keratin extract from human stratum corneum
Formulation PBS with 0.05% BSA. Protein G purified.
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23434601

 
novus biologicals canada
NBP23434601ML
0.1mL Each for $139.00

NBP23434605

 
novus biologicals canada
NBP23434605ML
0.5mL Each for $321.00
Description & Specifications

Specifications

Antigen Cytokeratin, Acidic (Type I or LMW)
Clone SPM115
Concentration 0.2mg/mL
Immunogen Solubilized keratin extract from human stratum corneum
Formulation PBS with 0.05% BSA. Protein G purified.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Bovine
Cross Reactivity Canine
Cross Reactivity Chicken
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Cross Reactivity Rabbit
Cross Reactivity Rat
Cross Reactivity Reptile
Isotype IgG1
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting