Please login to your online account to display your discounted pricing

Mouse anti-Cytokeratin, Acidic (Type I or LMW), Clone: SPM115, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23434605ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23434605

  • / Each
 

Description & Specifications

Specifications

Antigen Cytokeratin, Acidic (Type I or LMW)
Formulation PBS with 0.05% BSA. Protein G purified.
Host Species Mouse
Isotype IgG1
Quantity 0.5mL
Monoclonal or Polyclonal Monoclonal
Conjugate Unlabeled
Cross Reactivity Bovine
Cross Reactivity Canine
Cross Reactivity Chicken
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Cross Reactivity Rabbit
Cross Reactivity Rat
Cross Reactivity Reptile
Format Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Purification Method Purified
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Clone SPM115
Concentration 0.2mg/mL
Immunogen Solubilized keratin extract from human stratum corneum
Test Specificity This MAb recognizes the 56.5kDa (CK10); 50kDa (CK14); 50kDa (CK15); 48kDa (CK16); 40kDa (CK19) keratins of the acidic (Type I or LMW) subfamily. The acidic keratins have molecular weights (MW) of 56.5, 55, 51, 50, 50', 48, 46, 45, and 40kDa. MAb SPM116 recognizes the 65-67, 64, 59, 58, 56, and 52kDa keratins of basic subfamily.