Please login to your online account to display your discounted pricing

Mouse anti-Cytokeratin, Acidic (Type I or LMW), Clone: SPM115, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23439301MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23439301

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Host Species Mouse
Primary or Secondary Primary
Conjugate Unlabeled
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Antigen Cytokeratin, Acidic (Type I or LMW)
Regulatory Status RUO
Cross Reactivity Bovine
Cross Reactivity Canine
Cross Reactivity Chicken
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Cross Reactivity Rabbit
Cross Reactivity Rat
Cross Reactivity Reptile
Quantity 0.1mg
Purification Method Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Format Purified
Monoclonal or Polyclonal Monoclonal
Isotype IgG1
Concentration 1mg/mL
Reconstitution Protein A purified
Clone SPM115
Test Specificity This MAb recognizes the 56.5kDa (CK10); 50kDa (CK14); 50kDa (CK15); 48kDa (CK16); 40kDa (CK19) keratins of the acidic (Type I or LMW) subfamily. The acidic keratins have molecular weights (MW) of 56.5, 55, 51, 50, 50', 48, 46, 45, and 40kDa. MAb SPM116 recognizes the 65-67, 64, 59, 58, 56, and 52kDa keratins of basic subfamily.
Immunogen Solubilized keratin extract from human stratum corneum