Please login to your online account to display your discounted pricing

Mouse anti-Cytokeratin, Acidic (Type I or LMW), Clone: SPM115, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23439302MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23439302

  • / Each
 

Description & Specifications

Specifications

Antigen Cytokeratin, Acidic (Type I or LMW)
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Clone SPM115
Concentration 1mg/mL
Conjugate Unlabeled
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Host Species Murine
Immunogen Solubilized keratin extract from human stratum corneum
Isotype IgG1
Purification Method Purified
Quantity 0.2mg
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Test Specificity This MAb recognizes the 56.5kDa (CK10); 50kDa (CK14); 50kDa (CK15); 48kDa (CK16); 40kDa (CK19) keratins of the acidic (Type I or LMW) subfamily. The acidic keratins have molecular weights (MW) of 56.5, 55, 51, 50, 50', 48, 46, 45, and 40kDa. MAb SPM116 recognizes the 65-67, 64, 59, 58, 56, and 52kDa keratins of basic subfamily.
Reconstitution Protein A purified
Target Species Bovine
Target Species Canine
Target Species Chicken
Target Species Human
Target Species Murine
Target Species Primate
Target Species Rabbit
Target Species Rat
Target Species Reptile