Please login to your online account to display your discounted pricing

Mouse anti-Cytokeratin 8, Clone: TS1, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23450102MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23450102

  • / Each
 

Description & Specifications

Specifications

Format Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Isotype IgG1
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 3856
Cross Reactivity Human
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Gene Accession No. P05787
Host Species Mouse
Immunogen Keratin preparation from a human carcinoma
Purification Method Purified
Quantity 0.2mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Does not react to Rat.
Reconstitution Protein A purified
Clone TS1
Concentration 1mg/mL
Conjugate Unlabeled
Antigen Cytokeratin 8