Please login to your online account to display your discounted pricing

Mouse anti-Cytokeratin 8, Clone: TS1, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23450101MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23450101

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Antigen Cytokeratin 8
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Host Species Mouse
Immunogen Keratin preparation from a human carcinoma
Isotype IgG1
Quantity 0.1mg
Primary or Secondary Primary
Test Specificity Does not react to Rat.
Reconstitution Protein A purified
Clone TS1
Cross Reactivity Human
Format Purified
Gene Accession No. P05787
Purification Method Purified
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Monoclonal or Polyclonal Monoclonal
Concentration 1mg/mL
Conjugate Unlabeled
Gene ID (Entrez) 3856