Please login to your online account to display your discounted pricing

Mouse anti-Cytokeratin 8, Clone: SPM538, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23437005ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23437005

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Human
Cross Reactivity Rat
Cross Reactivity Zebrafish
Immunogen Cytoskeleton preparation containing cytokeratin 8
Format Purified
Host Species Mouse
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Formulation PBS with 0.05% BSA. Protein G purified.
Concentration 0.2mg/mL
Conjugate Unlabeled
Regulatory Status RUO
Gene ID (Entrez) 3856
Purification Method Purified
Isotype IgG1
Gene Accession No. P05787
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Clone SPM538
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Antigen Cytokeratin 8
Quantity 0.5mL