Please login to your online account to display your discounted pricing

Mouse anti-Cytokeratin 8, Clone: SPM538, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23437001ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23437001

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Human
Cross Reactivity Rat
Cross Reactivity Zebrafish
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Antigen Cytokeratin 8
Gene Accession No. P05787
Host Species Mouse
Immunogen Cytoskeleton preparation containing cytokeratin 8
Format Purified
Clone SPM538
Quantity 0.1mL
Conjugate Unlabeled
Formulation PBS with 0.05% BSA. Protein G purified.
Primary or Secondary Primary
Concentration 0.2mg/mL
Isotype IgG1
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 3856
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Purification Method Purified
Regulatory Status RUO