missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mouse anti-Cytokeratin 8, Clone: SPM538, Azide Free, Novus Biologicals™

$396.83 - $527.00

Specifications

Antigen Cytokeratin 8
Clone SPM538
Concentration 1mg/mL
Immunogen Cytoskeleton preparation containing cytokeratin 8
Gene Accession No. P05787
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP23441701 Novus Biologicals Canada
NBP23441701MG
0.1mg Each for $396.83
NBP23441702 Novus Biologicals Canada
NBP23441702MG
0.2mg Each for $527.00
Specifications Description & Specifications

Specifications

Cytokeratin 8
1mg/mL
P05787
10mM PBS. Does not contain BSA. Azide Free.
Primary
Murine
Unlabeled
Purified
SPM538
Cytoskeleton preparation containing cytokeratin 8
3856
Monoclonal
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG1
Purified
ELISA,Flow Cytometry,Immunocytochemistry,Immunofluorescence,Immunohistochemistry (Frozen),Immunohistochemistry (Paraffin),Immunoprecipitation,Western Blot