Please login to your online account to display your discounted pricing

Mouse anti-Cytokeratin 8, Clone: SPM538, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23441701MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23441701

  • / Each
 

Description & Specifications

Specifications

Format Purified
Conjugate Unlabeled
Gene Accession No. P05787
Quantity 0.1mg
Clone SPM538
Antigen Cytokeratin 8
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Regulatory Status RUO
Cross Reactivity Human
Cross Reactivity Rat
Cross Reactivity Zebrafish
Gene ID (Entrez) 3856
Purification Method Purified
Isotype IgG1
Primary or Secondary Primary
Reconstitution Protein A purified
Host Species Mouse
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Immunogen Cytoskeleton preparation containing cytokeratin 8
Monoclonal or Polyclonal Monoclonal
Concentration 1mg/mL