Please login to your online account to display your discounted pricing

Mouse anti-Cytokeratin 8, Clone: SPM192, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23435205ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23435205

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Host Species Mouse
Immunogen Keratin preparation from a human carcinoma
Gene ID (Entrez) 3856
Cross Reactivity Adenovirus
Cross Reactivity Human
Antigen Cytokeratin 8
Format Purified
Formulation PBS with 0.05% BSA. Protein G purified.
Gene Accession No. P05787
Isotype IgG1
Purification Method Purified
Quantity 0.5mL
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Test Specificity Anti-CK8 does not react with skeletal muscle or nerve cells. Does not react with Rat.
Clone SPM192
Concentration 0.2mg/mL
Conjugate Unlabeled