Please login to your online account to display your discounted pricing

Mouse anti-Cytokeratin 8, Clone: SPM192, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23440002MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23440002

  • / Each
 

Description & Specifications

Specifications

Antigen Cytokeratin 8
Host Species Mouse
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Gene ID (Entrez) 3856
Reconstitution Protein A purified
Conjugate Unlabeled
Immunogen Keratin preparation from a human carcinoma
Quantity 0.2mg
Cross Reactivity Human
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Gene Accession No. P05787
Isotype IgG1
Purification Method Purified
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Test Specificity Anti-CK8 does not react with skeletal muscle or nerve cells. Does not react with Rat.
Clone SPM192
Concentration 1mg/mL