Please login to your online account to display your discounted pricing

Mouse anti-Cytokeratin 8, Clone: SPM192, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23440001MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23440001

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Immunogen Keratin preparation from a human carcinoma
Quantity 0.1mg
Conjugate Unlabeled
Cross Reactivity Human
Antigen Cytokeratin 8
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Host Species Mouse
Purification Method Purified
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 3856
Test Specificity Anti-CK8 does not react with skeletal muscle or nerve cells. Does not react with Rat.
Reconstitution Protein A purified
Concentration 1mg/mL
Gene Accession No. P05787
Isotype IgG1
Regulatory Status RUO
Clone SPM192