Please login to your online account to display your discounted pricing

Mouse anti-Cytokeratin 8, Clone: K8.8, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23426901ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23426901

  • / Each
 

Description & Specifications

Specifications

Formulation PBS with 0.05% BSA. Protein G purified.
Cross Reactivity Human
Antigen Cytokeratin 8
Format Purified
Gene Accession No. P05787
Host Species Mouse
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Immunogen Keratin preparation from a human carcinoma
Isotype IgG1
Purification Method Purified
Quantity 0.1mL
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 3856
Clone K8.8
Concentration 0.2mg/mL
Conjugate Unlabeled
Test Specificity Anti-CK8 does not react with skeletal muscle or nerve cells. Epithelioid sarcoma, chordoma, and adamantinoma show strong positivity corresponding to that of simple epithelia (with antibodies against CK8, CK18 and CK19). Reportedly, anti-CK8 is useful for the differentiation of lobular (ring-like, perinuclear) from ductal (peripheral-predominant) carcinoma of the breast. Does not react with Rat.