Please login to your online account to display your discounted pricing

Mouse anti-Cytokeratin 8, Clone: K8/383, Novus Biologicals™

$139.00 - $321.00

Specifications

Antigen Cytokeratin 8
Clone K8/383
Concentration 0.2mg/mL
Immunogen Recombinant human cytokeratin 8 protein
Gene Accession No. P05787
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23426601

 
novus biologicals canada
NBP23426601ML
0.1mL Each for $139.00

NBP23426605

 
novus biologicals canada
NBP23426605ML
0.5mL Each for $321.00
Description & Specifications

Specifications

Antigen Cytokeratin 8
Clone K8/383
Concentration 0.2mg/mL
Immunogen Recombinant human cytokeratin 8 protein
Gene Accession No. P05787
Gene ID (Entrez) 3856
Formulation PBS with 0.05% BSA. Protein G purified.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Human
Cross Reactivity Rat
Cross Reactivity Zebrafish
Isotype IgG1
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting