Please login to your online account to display your discounted pricing

Mouse anti-Cytokeratin 8, Clone: K8/383, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23426601ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23426601

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Format Purified
Formulation PBS with 0.05% BSA. Protein G purified.
Host Species Mouse
Isotype IgG1
Primary or Secondary Primary
Clone K8/383
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Rat
Cross Reactivity Zebrafish
Antigen Cytokeratin 8
Gene Accession No. P05787
Immunogen Recombinant human cytokeratin 8 protein
Purification Method Purified
Quantity 0.1mL
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Gene ID (Entrez) 3856
Concentration 0.2mg/mL
Monoclonal or Polyclonal Monoclonal