Please login to your online account to display your discounted pricing

Mouse anti-Cytokeratin 8, Clone: K8/383, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23426605ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23426605

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Monoclonal or Polyclonal Monoclonal
Immunogen Recombinant human cytokeratin 8 protein
Primary or Secondary Primary
Gene ID (Entrez) 3856
Quantity 0.5mL
Isotype IgG1
Regulatory Status RUO
Host Species Mouse
Clone K8/383
Cross Reactivity Human
Cross Reactivity Rat
Cross Reactivity Zebrafish
Antigen Cytokeratin 8
Concentration 0.2mg/mL
Format Purified
Conjugate Unlabeled
Formulation PBS with 0.05% BSA. Protein G purified.
Gene Accession No. P05787
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Purification Method Purified