missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mouse anti-Cytokeratin 8, Clone: K8/383, Azide Free, Novus Biologicals™

$384.00 - $527.00

Specifications

Antigen Cytokeratin 8
Clone K8/383
Concentration 1mg/mL
Immunogen Recombinant human cytokeratin 8 protein
Gene Accession No. P05787
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP23465601 Novus Biologicals Canada
NBP23465601MG
0.1mg Each for $384.00
NBP23465602 Novus Biologicals Canada
NBP23465602MG
0.2mg Each for $527.00
Specifications Description & Specifications

Specifications

Cytokeratin 8
1mg/mL
P05787
10mM PBS. Does not contain BSA. Azide Free.
Primary
Murine
Unlabeled
Purified
K8/383
Recombinant human cytokeratin 8 protein
3856
Monoclonal
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG1
Purified
ELISA,Flow Cytometry,Immunocytochemistry,Immunofluorescence,Immunohistochemistry (Frozen),Immunohistochemistry (Paraffin),Immunoprecipitation,Western Blot