Please login to your online account to display your discounted pricing

Mouse anti-Cytokeratin 8, Clone: K8/383, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23465601MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23465601

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Antigen Cytokeratin 8
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Gene Accession No. P05787
Host Species Mouse
Immunogen Recombinant human cytokeratin 8 protein
Isotype IgG1
Quantity 0.1mg
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 3856
Reconstitution Protein A purified
Concentration 1mg/mL
Cross Reactivity Human
Cross Reactivity Rat
Cross Reactivity Zebrafish
Purification Method Purified
Regulatory Status RUO
Primary or Secondary Primary
Clone K8/383
Conjugate Unlabeled