Please login to your online account to display your discounted pricing

Mouse anti-Cytokeratin 8, Clone: H1 + TS1, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23426805ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23426805

  • / Each
 

Description & Specifications

Specifications

Regulatory Status RUO
Host Species Mouse
Gene ID (Entrez) 3856
Clone H1 + TS1
Isotype IgG1
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Cross Reactivity Human
Cross Reactivity Rat
Cross Reactivity Zebrafish
Conjugate Unlabeled
Format Purified
Immunogen Cytoskeleton preparation containing cytokeratin 8 (H1); Keratin preparation from a human carcinoma (TS1)
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Purification Method Purified
Quantity 0.5mL
Reconstitution Protein A purified
Antigen Cytokeratin 8
Primary or Secondary Primary
Formulation PBS with 0.05% BSA.
Monoclonal or Polyclonal Monoclonal
Concentration 0.2mg/mL
Gene Accession No. P05787