Please login to your online account to display your discounted pricing

Mouse anti-Cytokeratin 8, Clone: H1 + TS1, Azide Free, Novus Biologicals™

$384.00 - $527.00

Specifications

Antigen Cytokeratin 8
Clone H1 + TS1
Concentration 1mg/mL
Immunogen Cytoskeleton preparation containing cytokeratin 8 (H1); Keratin preparation from a human carcinoma (TS1)
Gene Accession No. P05787
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23462701

 
novus biologicals canada
NBP23462701MG
0.1mg Each for $384.00

NBP23462702

 
novus biologicals canada
NBP23462702MG
0.2mg Each for $527.00
Description & Specifications

Specifications

Antigen Cytokeratin 8
Clone H1 + TS1
Concentration 1mg/mL
Immunogen Cytoskeleton preparation containing cytokeratin 8 (H1); Keratin preparation from a human carcinoma (TS1)
Gene Accession No. P05787
Gene ID (Entrez) 3856
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Human
Cross Reactivity Rat
Cross Reactivity Zebrafish
Isotype IgG1
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting