Please login to your online account to display your discounted pricing

Mouse anti-Cytokeratin 8, Clone: H1 + TS1, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23462701MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23462701

  • / Each
 

Description & Specifications

Specifications

Host Species Mouse
Immunogen Cytoskeleton preparation containing cytokeratin 8 (H1); Keratin preparation from a human carcinoma (TS1)
Primary or Secondary Primary
Format Purified
Isotype IgG1
Antigen Cytokeratin 8
Monoclonal or Polyclonal Monoclonal
Concentration 1mg/mL
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Cross Reactivity Human
Cross Reactivity Rat
Cross Reactivity Zebrafish
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Quantity 0.1mg
Purification Method Purified
Gene ID (Entrez) 3856
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Clone H1 + TS1
Gene Accession No. P05787
Reconstitution Protein A purified
Conjugate Unlabeled