Please login to your online account to display your discounted pricing

Mouse anti-Cytokeratin 8, Clone: H1, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23462602MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23462602

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Immunogen Cytoskeleton preparation containing cytokeratin 8
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Rat
Cross Reactivity Zebrafish
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Gene Accession No. P05787
Isotype IgG1
Purification Method Purified
Quantity 0.2mg
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 3856
Reconstitution Protein A purified
Clone H1
Antigen Cytokeratin 8
Format Purified
Host Species Mouse
Regulatory Status RUO
Concentration 1mg/mL