Please login to your online account to display your discounted pricing

Mouse anti-Cytokeratin 8, Clone: H1, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23462601MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23462601

  • / Each
 

Description & Specifications

Specifications

Antigen Cytokeratin 8
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blot
Clone H1
Concentration 1mg/mL
Conjugate Unlabeled
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Gene Accession No. P05787
Host Species Murine
Immunogen Cytoskeleton preparation containing cytokeratin 8
Isotype IgG1
Purification Method Purified
Quantity 0.1mg
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 3856
Reconstitution Protein A purified
Target Species Human
Target Species Rat
Target Species Zebrafish