Please login to your online account to display your discounted pricing

Mouse anti-Cytokeratin 8/18, Clone: K8.8 + DC10, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23434401ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23434401

  • / Each
 

Description & Specifications

Specifications

Antigen Cytokeratin 8/18
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Clone K8.8 + DC10
Concentration 0.2mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Formulation PBS with 0.05% BSA.
Gene Accession No. P05787
Host Species Mouse
Immunogen Keratin preparation from a human carcinoma (K8.8); PMC-42 human breast carcinoma cells (DC10)
Isotype IgG1, κ
Purification Method Purified
Quantity 0.1mL
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 3856
Reconstitution Protein A purified