Please login to your online account to display your discounted pricing

Mouse anti-Cytokeratin 8/18, Clone: K8.8 + DC10, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23465501MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23465501

  • / Each
 

Description & Specifications

Specifications

Antigen Cytokeratin 8/18
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Gene Accession No. P05787
Host Species Mouse
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Immunogen Keratin preparation from a human carcinoma (K8.8); PMC-42 human breast carcinoma cells (DC10)
Isotype IgG1, κ
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 3856
Reconstitution Protein A purified
Clone K8.8 + DC10
Concentration 1mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Purification Method Purified
Quantity 0.1mg
Primary or Secondary Primary