Please login to your online account to display your discounted pricing

Mouse anti-Cytokeratin 6, Clone: SPM269, Azide Free, Novus Biologicals™

$384.00 - $676.00

Specifications

Antigen Cytokeratin 6
Clone SPM269
Concentration 1mg/mL
Immunogen A synthetic peptide of 11 amino acids (GSSTIKYTTTS) from C-terminus of human keratin 6
Gene ID (Entrez) 3853
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23440601

 
novus biologicals canada
NBP23440601MG
0.1mg Each for $384.00

NBP23440602

 
novus biologicals canada
NBP23440602MG
0.2mg Each for $676.00
Description & Specifications

Specifications

Antigen Cytokeratin 6
Clone SPM269
Concentration 1mg/mL
Immunogen A synthetic peptide of 11 amino acids (GSSTIKYTTTS) from C-terminus of human keratin 6
Gene ID (Entrez) 3853
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Murine
Isotype IgG2a, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blot