Please login to your online account to display your discounted pricing

Mouse anti-Cytokeratin 6, Clone: SPM269, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23440601MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23440601

  • / Each
 

Description & Specifications

Specifications

Format Purified
Cross Reactivity Human
Cross Reactivity Mouse
Isotype IgG2a, κ
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Antigen Cytokeratin 6
Quantity 0.1mg
Concentration 1mg/mL
Regulatory Status RUO
Primary or Secondary Primary
Host Species Mouse
Clone SPM269
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Conjugate Unlabeled
Purification Method Purified
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 3853
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Test Specificity This MAb recognizes a protein of 56kDa, identified as cytokeratin 6 (CK6).
Reconstitution Protein A purified
Immunogen A synthetic peptide of 11 amino acids (GSSTIKYTTTS) from C-terminus of human keratin 6