Please login to your online account to display your discounted pricing

Mouse anti-Cytokeratin 6, Clone: SPM269, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23440602MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23440602

  • / Each
 

Description & Specifications

Specifications

Format Purified
Host Species Mouse
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Immunogen A synthetic peptide of 11 amino acids (GSSTIKYTTTS) from C-terminus of human keratin 6
Purification Method Purified
Quantity 0.2mg
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Antigen Cytokeratin 6
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Isotype IgG2a, κ
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 3853
Test Specificity This MAb recognizes a protein of 56kDa, identified as cytokeratin 6 (CK6).
Reconstitution Protein A purified
Clone SPM269
Concentration 1mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse