Please login to your online account to display your discounted pricing

Mouse anti-Cytokeratin 6, Clone: LHK6, Novus Biologicals™

$139.00 - $321.00

Specifications

Antigen Cytokeratin 6
Clone LHK6
Concentration 0.2mg/mL
Immunogen A synthetic peptide of 11 amino acids (GSSTIKYTTTS) from C-terminus of human keratin 6
Gene Accession No. P02538
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23423201

 
novus biologicals canada
NBP23423201ML
0.1mL Each for $139.00

NBP23423205

 
novus biologicals canada
NBP23423205ML
0.5mL Each for $321.00
Description & Specifications

Specifications

Antigen Cytokeratin 6
Clone LHK6
Concentration 0.2mg/mL
Immunogen A synthetic peptide of 11 amino acids (GSSTIKYTTTS) from C-terminus of human keratin 6
Gene Accession No. P02538
Gene ID (Entrez) 3853
Formulation PBS with 0.05% BSA.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Human
Cross Reactivity Mouse
Isotype IgG2a, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting