Please login to your online account to display your discounted pricing

Mouse anti-Cytokeratin 6, Clone: LHK6, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23423201ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23423201

  • / Each
 

Description & Specifications

Specifications

Antigen Cytokeratin 6
Format Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Cross Reactivity Human
Cross Reactivity Mouse
Formulation PBS with 0.05% BSA.
Gene Accession No. P02538
Host Species Mouse
Immunogen A synthetic peptide of 11 amino acids (GSSTIKYTTTS) from C-terminus of human keratin 6
Isotype IgG2a, κ
Purification Method Purified
Quantity 0.1mL
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 3853
Reconstitution Protein A purified
Clone LHK6
Concentration 0.2mg/mL
Conjugate Unlabeled
Primary or Secondary Primary