Please login to your online account to display your discounted pricing

Mouse anti-Cytokeratin 6, Clone: LHK6, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23466901MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23466901

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Cross Reactivity Human
Cross Reactivity Mouse
Antigen Cytokeratin 6
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Gene Accession No. P02538
Host Species Mouse
Immunogen A synthetic peptide of 11 amino acids (GSSTIKYTTTS) from C-terminus of human keratin 6
Isotype IgG2a, κ
Quantity 0.1mg
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 3853
Clone LHK6
Concentration 1mg/mL
Conjugate Unlabeled
Purification Method Purified
Regulatory Status RUO
Reconstitution Protein A purified