Please login to your online account to display your discounted pricing

Mouse anti-Cytokeratin 5, Clone: 10C11E6, Novus Biologicals™

$367.12 - $397.00

Specifications

Antigen Cytokeratin 5
Clone 10C11E6
Concentration 1mg/mL
Immunogen Purified recombinant fragment of human CK5 (AA: 316-590) expressed in E. Coli.
Gene ID (Entrez) 3852
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP237334

 
novus biologicals canada
NBP237334
0.1mL Each for $367.12

NBP237400

 
novus biologicals canada
NBP237400
0.1mL Each for $397.00
Description & Specifications

Specifications

Antigen Cytokeratin 5
Clone 10C11E6
Concentration 1mg/mL
Immunogen Purified recombinant fragment of human CK5 (AA: 316-590) expressed in E. Coli.
Gene ID (Entrez) 3852
Formulation PBS
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Human
Isotype IgG1
Conjugate Unlabeled
Format Affinity Purified
Purification Method Affinity Purified
Applications ELISA
Applications Immunohistochemistry
Applications Western Blotting