Please login to your online account to display your discounted pricing

Mouse anti-Cytokeratin 5, Clone: 10C11E6, Novus Biologicals™

Manufacturer: novus biologicals canada  NBP237400

 View all options for this product


For Research Use Only

Catalog No. NBP237400


Description & Specifications


Antigen Cytokeratin 5
Applications ELISA
Applications Immunohistochemistry
Applications Western Blotting
Clone 10C11E6
Concentration 1mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Affinity Purified
Formulation PBS
Host Species Mouse
Immunogen Purified recombinant fragment of human CK5 (AA: 316-590) expressed in E. Coli.
Isotype IgG1
Purification Method Affinity Purified
Quantity 0.1mL
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 3852