Please login to your online account to display your discounted pricing

Mouse anti-Cytokeratin 19, Clone: SPM561, Novus Biologicals™

$139.00 - $321.00

Specifications

Antigen Cytokeratin 19
Clone SPM561
Concentration 0.2mg/mL
Immunogen Human mammary epithelial organoids
Gene Accession No. P08727
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23437901

 
novus biologicals canada
NBP23437901ML
0.1mL Each for $139.00

NBP23437905

 
novus biologicals canada
NBP23437905ML
0.5mL Each for $321.00
Description & Specifications

Specifications

Antigen Cytokeratin 19
Clone SPM561
Concentration 0.2mg/mL
Immunogen Human mammary epithelial organoids
Gene Accession No. P08727
Gene ID (Entrez) 3880
Formulation PBS with 0.05% BSA.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Human
Cross Reactivity Mouse
Isotype IgG1, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting