Please login to your online account to display your discounted pricing

Mouse anti-Cytokeratin 19, Clone: SPM561, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23437901ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23437901

  • / Each
 

Description & Specifications

Specifications

Formulation PBS with 0.05% BSA.
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Immunogen Human mammary epithelial organoids
Isotype IgG1, κ
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 3880
Conjugate Unlabeled
Antigen Cytokeratin 19
Format Purified
Gene Accession No. P08727
Host Species Mouse
Purification Method Purified
Quantity 0.1mL
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Test Specificity This Ab reacts with the rod domain of human cytokeratin 19 (CK19), a polypeptide of 40kDa. Anti-CK19 is a useful marker for detection of tumor cells in lymph nodes, peripheral blood, bone marrow and breast cancer.
Reconstitution Protein A purified
Clone SPM561
Concentration 0.2mg/mL
Cross Reactivity Human
Cross Reactivity Mouse