Please login to your online account to display your discounted pricing

Mouse anti-Cytokeratin 19, Clone: SPM561, Azide Free, Novus Biologicals™

$396.83 - $527.00

Specifications

Antigen Cytokeratin 19
Clone SPM561
Concentration 1mg/mL
Immunogen Human mammary epithelial organoids
Gene Accession No. P08727
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23442601

 
novus biologicals canada
NBP23442601MG
0.1mg Each for $396.83

NBP23442602

 
novus biologicals canada
NBP23442602MG
0.2mg Each for $527.00
Description & Specifications

Specifications

Antigen Cytokeratin 19
Clone SPM561
Concentration 1mg/mL
Immunogen Human mammary epithelial organoids
Gene Accession No. P08727
Gene ID (Entrez) 3880
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Human
Cross Reactivity Mouse
Isotype IgG1, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting