Please login to your online account to display your discounted pricing

Mouse anti-Cytokeratin 19, Clone: SPM561, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23442601MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23442601

  • / Each
 

Description & Specifications

Specifications

Immunogen Human mammary epithelial organoids
Antigen Cytokeratin 19
Regulatory Status RUO
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Gene ID (Entrez) 3880
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Clone SPM561
Isotype IgG1, κ
Gene Accession No. P08727
Quantity 0.1mg
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Test Specificity This Ab reacts with the rod domain of human cytokeratin 19 (CK19), a polypeptide of 40kDa. Anti-CK19 is a useful marker for detection of tumor cells in lymph nodes, peripheral blood, bone marrow and breast cancer.
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Monoclonal or Polyclonal Monoclonal
Concentration 1mg/mL
Reconstitution Protein A purified
Purification Method Purified
Format Purified
Primary or Secondary Primary