Please login to your online account to display your discounted pricing

Mouse anti-Cytokeratin 19, Clone: SPM266, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23435705ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23435705

  • / Each
 

Description & Specifications

Specifications

Quantity 0.5mL
Cross Reactivity Human
Host Species Mouse
Monoclonal or Polyclonal Monoclonal
Formulation PBS with 0.05% BSA.
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Isotype IgG2a, κ
Gene Accession No. P08727
Primary or Secondary Primary
Clone SPM266
Conjugate Unlabeled
Antigen Cytokeratin 19
Gene ID (Entrez) 3880
Immunogen Human breast cancer MCF-7 cells
Format Purified
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Test Specificity This MAb reacts with the rod domain of human cytokeratin-19 (CK19), a polypeptide of 40kDa. Its epitope maps between amino acid 312-335.
Concentration 0.2mg/mL
Reconstitution Protein A purified
Purification Method Purified
Regulatory Status RUO