Please login to your online account to display your discounted pricing

Mouse anti-Cytokeratin 19, Clone: SPM266, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23440501MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23440501

  • / Each
 

Description & Specifications

Specifications

Format Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Conjugate Unlabeled
Cross Reactivity Human
Antigen Cytokeratin 19
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Gene Accession No. P08727
Host Species Mouse
Immunogen Human breast cancer MCF-7 cells
Isotype IgG2a, κ
Purification Method Purified
Quantity 0.1mg
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 3880
Test Specificity This MAb reacts with the rod domain of human cytokeratin-19 (CK19), a polypeptide of 40kDa. Its epitope maps between amino acid 312-335.
Reconstitution Protein A purified
Clone SPM266
Concentration 1mg/mL