Please login to your online account to display your discounted pricing

Mouse anti-Cytokeratin 19, Clone: SPM266, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23440502MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23440502

  • / Each
 

Description & Specifications

Specifications

Formulation 10mM PBS. Does not contain BSA. Azide Free.
Clone SPM266
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Primary or Secondary Primary
Quantity 0.2mg
Format Purified
Concentration 1mg/mL
Isotype IgG2a, κ
Host Species Mouse
Gene Accession No. P08727
Purification Method Purified
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Regulatory Status RUO
Gene ID (Entrez) 3880
Reconstitution Protein A purified
Cross Reactivity Human
Antigen Cytokeratin 19
Monoclonal or Polyclonal Monoclonal
Test Specificity This MAb reacts with the rod domain of human cytokeratin-19 (CK19), a polypeptide of 40kDa. Its epitope maps between amino acid 312-335.
Immunogen Human breast cancer MCF-7 cells
Conjugate Unlabeled