Please login to your online account to display your discounted pricing

Mouse anti-Cytokeratin 19, Clone: BA17, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23427105ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23427105

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Quantity 0.5mL
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Monoclonal or Polyclonal Monoclonal
Cross Reactivity Human
Cross Reactivity Mouse
Antigen Cytokeratin 19
Format Purified
Gene Accession No. P08727
Host Species Mouse
Immunogen Human mammary epithelial organoids
Isotype IgG1, κ
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3880
Concentration 0.2mg/mL
Conjugate Unlabeled
Formulation PBS with 0.05% BSA.
Purification Method Purified
Reconstitution Protein A purified
Clone BA17