missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mouse anti-Cytokeratin 19, Clone: BA17, Azide Free, Novus Biologicals™

$384.00 - $676.00

Specifications

Antigen Cytokeratin 19
Clone BA17
Concentration 1mg/mL
Immunogen Human mammary epithelial organoids
Gene Accession No. P08727
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP23456001 Novus Biologicals Canada
NBP23456001MG
0.1mg Each for $384.00
NBP23456002 Novus Biologicals Canada
NBP23456002MG
0.2mg Each for $676.00
Specifications Description & Specifications

Specifications

Cytokeratin 19
1mg/mL
P08727
10mM PBS. Does not contain BSA. Azide Free.
Primary
Murine
Unlabeled
Purified
BA17
Human mammary epithelial organoids
3880
Monoclonal
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG1, κ
Purified
ELISA,Flow Cytometry,Immunocytochemistry,Immunofluorescence,Immunohistochemistry (Frozen),Immunohistochemistry (Paraffin),Immunoprecipitation,Western Blot