Please login to your online account to display your discounted pricing

Mouse anti-Cytokeratin 19, Clone: A53-B/A2.26, Novus Biologicals™

$329.00 - $472.00

Specifications

Antigen Cytokeratin 19
Clone A53-B/A2.26
Concentration 0.2mg/mL
Immunogen Human breast cancer MCF-7 cells were used as the immunogen for the Keratin-19 antibody.
Gene Accession No. P08727
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP21518601

 
novus biologicals canada
NBP21518601MG
0.1mg Each for $329.00

NBP21518602

 
novus biologicals canada
NBP21518602MG
0.2mg Each for $472.00
Description & Specifications

Specifications

Antigen Cytokeratin 19
Clone A53-B/A2.26
Concentration 0.2mg/mL
Immunogen Human breast cancer MCF-7 cells were used as the immunogen for the Keratin-19 antibody.
Gene Accession No. P08727
Gene ID (Entrez) 3880
Formulation PBS with 0.05% BSA. Protein G purified.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Human
Isotype IgG2a, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting