Please login to your online account to display your discounted pricing

Mouse anti-Cytokeratin 19, Clone: A53-B/A2.26 + BA17, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23427205ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23427205

  • / Each
 

Description & Specifications

Specifications

Format Purified
Formulation PBS with 0.05% BSA.
Gene Accession No. P08727
Host Species Mouse
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Immunogen Human breast cancer MCF-7 cells (A53-B/A2); Human mammary epithelial organoids (BA17)
Isotype IgG1, κ
Purification Method Purified
Quantity 0.5mL
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Antigen Cytokeratin 19
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 3880
Reconstitution Protein A purified
Clone A53-B/A2.26 + BA17
Concentration 0.2mg/mL