Please login to your online account to display your discounted pricing

Mouse anti-Cytokeratin 19, Clone: A53-B/A2.26 + BA17, Novus Biologicals™

Manufacturer: novus biologicals canada  NBP23427201ML

 View all options for this product


For Research Use Only

Catalog No. NBP23427201


Description & Specifications


Antigen Cytokeratin 19
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Clone A53-B/A2.26 + BA17
Concentration 0.2mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Format Purified
Formulation PBS with 0.05% BSA.
Gene Accession No. P08727
Host Species Mouse
Immunogen Human breast cancer MCF-7 cells (A53-B/A2); Human mammary epithelial organoids (BA17)
Isotype IgG1, κ
Purification Method Purified
Quantity 0.1mL
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 3880
Reconstitution Protein A purified