Please login to your online account to display your discounted pricing

Mouse anti-Cytokeratin 19, Clone: A53-B/A2.26 + BA17, Azide Free, Novus Biologicals™

$384.00 - $527.00

Specifications

Antigen Cytokeratin 19
Clone A53-B/A2.26 + BA17
Concentration 1mg/mL
Immunogen Human breast cancer MCF-7 cells (A53-B/A2); Human mammary epithelial organoids (BA17)
Gene Accession No. P08727
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23454801

 
novus biologicals canada
NBP23454801MG
0.1mg Each for $384.00

NBP23454802

 
novus biologicals canada
NBP23454802MG
0.2mg Each for $527.00
Description & Specifications

Specifications

Antigen Cytokeratin 19
Clone A53-B/A2.26 + BA17
Concentration 1mg/mL
Immunogen Human breast cancer MCF-7 cells (A53-B/A2); Human mammary epithelial organoids (BA17)
Gene Accession No. P08727
Gene ID (Entrez) 3880
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Murine
Isotype IgG1, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blot