Please login to your online account to display your discounted pricing

Mouse anti-Cytokeratin 19, Clone: A53-B/A2.26 + BA17, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23454801MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23454801

  • / Each
 

Description & Specifications

Specifications

Format Purified
Immunogen Human breast cancer MCF-7 cells (A53-B/A2); Human mammary epithelial organoids (BA17)
Isotype IgG1, κ
Conjugate Unlabeled
Gene ID (Entrez) 3880
Cross Reactivity Human
Cross Reactivity Mouse
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Antigen Cytokeratin 19
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Monoclonal or Polyclonal Monoclonal
Regulatory Status RUO
Concentration 1mg/mL
Clone A53-B/A2.26 + BA17
Gene Accession No. P08727
Reconstitution Protein A purified
Quantity 0.1mg
Host Species Mouse
Purification Method Purified
Primary or Secondary Primary
Formulation 10mM PBS. Does not contain BSA. Azide Free.