Please login to your online account to display your discounted pricing

Mouse anti-Cytokeratin 18, Clone: SPM510, Novus Biologicals™

Manufacturer: novus biologicals canada  NBP23436701ML

 View all options for this product


For Research Use Only

Catalog No. NBP23436701


Description & Specifications


Antigen Cytokeratin 18
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Clone SPM510
Concentration 0.2mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Formulation PBS with 0.05% BSA.
Gene Accession No. P05783
Host Species Mouse
Immunogen Human breast cancer PMC 42 cells
Isotype IgG1, κ
Purification Method Purified
Quantity 0.1mL
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 3875
Test Specificity This MAb reacts with a wide variety of simple epithelia. It does not react with stratified squamous epithelia. It reacts with epithelial tumors of the gastrointestinal tract, lung, breast, pancreas, ovary, and thyroid. Does not react with Bovine, Canine, Hamster, Mouse, Porcine, Rat or Sheep.
Reconstitution Protein A purified